ENA423200716 Enamine

4-[(oxiran-2-yl)methyl]thiomorpholine

  • MDL number MFCD11172213
  • Cas Number : 1042782-25-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-160604
Availability details for
Quantity
  • Availability
  • Add to Cart