ENA423200723 Enamine

ethyl 2-[(propan-2-yl)amino]butanoate

  • MDL number MFCD12150510
  • Cas Number : 1218559-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160654

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160654
Availability details for
Quantity
  • Availability
  • Add to Cart