ENA423200751 Enamine

4-fluoro-N-(pyrimidin-5-ylmethyl)aniline

  • MDL number MFCD16864885
  • Cas Number : 1339776-07-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160691

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160691
Availability details for
Quantity
  • Availability
  • Add to Cart