ENA423200817 Enamine

2-fluoro-N-(1,3-thiazol-5-ylmethyl)aniline

  • MDL number MFCD16865392
  • Cas Number : 1341402-11-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160757

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160757
Availability details for
Quantity
  • Availability
  • Add to Cart