ENA423200899 Enamine

4-fluoro-N-(1H-imidazol-2-ylmethyl)-3-methylaniline

  • MDL number MFCD17227000
  • Cas Number : 1338981-89-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-160841

Safety Information

msds https://www.enaminestore.com/catalog/EN300-160841
Availability details for
Quantity
  • Availability
  • Add to Cart