ENA423201062 Enamine

N-(2,5-dimethylphenyl)thian-3-amine

  • MDL number MFCD17238272
  • Cas Number : 1341051-81-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-161004

Safety Information

msds https://www.enaminestore.com/catalog/EN300-161004
Availability details for
Quantity
  • Availability
  • Add to Cart