ENA423201201 Enamine

2-{[(3-ethoxyphenyl)methyl]amino}ethan-1-ol

  • MDL number MFCD07405614
  • Cas Number : 889949-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-161148

Safety Information

msds https://www.enaminestore.com/catalog/EN300-161148
Availability details for
Quantity
  • Availability
  • Add to Cart