ENA423201372 Enamine

3-bromo-4-methyl-N-(propan-2-yl)aniline

  • MDL number MFCD11938420
  • Cas Number : 298185-50-9
  • Molecular Weight : 228.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-161320

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-161320
Availability details for
Quantity
  • Availability
  • Add to Cart