ENA423201727 Enamine

2-methyl-N-[1-(1,3-thiazol-2-yl)ethyl]cyclohexan-1-amine

  • MDL number MFCD20312553
  • Cas Number : 1338959-85-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-161709

Safety Information

msds https://www.enaminestore.com/catalog/EN300-161709
Availability details for
Quantity
  • Availability
  • Add to Cart