ENA423202226 Enamine

N-ethyl-5-methyl-2-nitroaniline

  • MDL number MFCD23725901
  • Cas Number : 502178-78-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-162215

Safety Information

msds https://www.enaminestore.com/catalog/EN300-162215
Availability details for
Quantity
  • Availability
  • Add to Cart