ENA423202314 Enamine

[(2-bromo-5-chlorophenyl)methyl](2-methylpropyl)amine

  • MDL number MFCD21705593
  • Cas Number : 1492317-53-9
  • Molecular Weight : 276.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-162317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-162317
Availability details for
Quantity
  • Availability
  • Add to Cart