ENA423202641 Enamine

(1-phenylethyl)(propan-2-yl)amine

  • MDL number MFCD00463431
  • Cas Number : 19302-16-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-162761

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-162761
Availability details for
Quantity
  • Availability
  • Add to Cart