ENA423203064 Enamine

N-(cyclopropylmethyl)-6-methylpyridin-3-amine

  • MDL number MFCD24119379
  • Cas Number : 1523513-70-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-163290

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-163290
Availability details for
Quantity
  • Availability
  • Add to Cart