ENA423203177 Enamine

[(1-ethyl-1H-pyrazol-5-yl)methyl](1-methoxypropan-2-yl)amine

  • MDL number MFCD27980798
  • Cas Number : 1602946-21-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-163406

Safety Information

msds https://www.enaminestore.com/catalog/EN300-163406
Availability details for
Quantity
  • Availability
  • Add to Cart