ENA423203678 Enamine

2-[(cyclopropylamino)methyl]phenol

  • MDL number MFCD09047227
  • Cas Number : 643007-91-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164098

Safety Information

msds https://www.enaminestore.com/catalog/EN300-164098
Availability details for
Quantity
  • Availability
  • Add to Cart