ENA423203725 Enamine

2-[(tert-butylamino)methyl]-4-chlorophenol

  • MDL number MFCD09043828
  • Cas Number : 926211-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164146

Safety Information

msds https://www.enaminestore.com/catalog/EN300-164146
Availability details for
Quantity
  • Availability
  • Add to Cart