ENA423203798 Enamine

4-ethyl-N-(propan-2-yl)aniline

  • MDL number MFCD11146067
  • Cas Number : 103093-97-6
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-164219

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164219
Availability details for
Quantity
  • Availability
  • Add to Cart