ENA423203817 Enamine

2-[2-(aminomethyl)phenoxy]ethan-1-amine dihydrochloride

  • MDL number MFCD27959469
  • Cas Number : 1803588-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164412

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164412
Availability details for
Quantity
  • Availability
  • Add to Cart