ENA423203818 Enamine

[(2-chlorophenyl)methyl](2-methylbutan-2-yl)amine

  • MDL number MFCD09688589
  • Cas Number : 931992-06-2
  • Molecular Weight : 211.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-164235

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164235
Availability details for
Quantity
  • Availability
  • Add to Cart