ENA423203819 Enamine

[(2,4-dichlorophenyl)methyl](2-methylbutan-2-yl)amine

  • MDL number MFCD09688435
  • Cas Number : 931991-22-9
  • Molecular Weight : 246.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-164236

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164236
Availability details for
Quantity
  • Availability
  • Add to Cart