ENA423203830 Enamine

2-methoxy-N-(propan-2-yl)aniline

  • Molecular Weight : 165.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-isopropyl-2-methoxyaniline
Inchi Code 1S/C10H15NO/c1-8(2)11-9-6-4-5-7-10(9)12-3/h4-8,11H,1-3H3
Inchi Key WFQXGJMHCGBRQJ-UHFFFAOYSA-N

Description

Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164247
Availability details for
Quantity
  • Availability
  • Add to Cart