ENA423203929 Enamine

3-[(cyclopropylamino)methyl]benzamide

  • MDL number MFCD09931108
  • Cas Number : 1016509-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164346

Safety Information

msds https://www.enaminestore.com/catalog/EN300-164346
Availability details for
Quantity
  • Availability
  • Add to Cart