ENA423203989 Enamine

1-(pyridin-2-yl)prop-2-yn-1-ol

  • MDL number MFCD18207399
  • Cas Number : 276884-47-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-164408
Availability details for
Quantity
  • Availability
  • Add to Cart