ENA423204024 Enamine

3-methyl-N-(pentan-3-yl)aniline

  • MDL number MFCD11138318
  • Cas Number : 1019587-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164443

Safety Information

msds https://www.enaminestore.com/catalog/EN300-164443
Availability details for
Quantity
  • Availability
  • Add to Cart