ENA423204058 Enamine

N-(3-methylbutan-2-yl)aniline

  • MDL number MFCD11138705
  • Cas Number : 100054-14-6
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-164477

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P312,P330,P363,P501
MSDS https://www.enaminestore.com/catalog/EN300-164477
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart