ENA423204308 Enamine

2-bromo-4-methyl-N-(propan-2-yl)aniline

  • MDL number MFCD11141242
  • Cas Number : 1019616-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-164727

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-164727
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart