ENA423204767 Enamine

4-methyl-N-(2-methylpropyl)aniline

  • MDL number MFCD11147194
  • Cas Number : 54837-93-3
  • Molecular Weight : 163.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-165187

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-165187
Availability details for
Quantity
  • Availability
  • Add to Cart