ENA423204789 Enamine

4-methoxy-N-(3-methylbutyl)aniline

  • MDL number MFCD11147319
  • Cas Number : 847224-60-6
  • Molecular Weight : 193.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-165209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-165209
Availability details for
Quantity
  • Availability
  • Add to Cart