ENA423204824 Enamine

2-[(cyclopropylmethyl)amino]-N,N-dimethylacetamide

  • MDL number MFCD11150453
  • Cas Number : 1021038-48-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-165246

Safety Information

msds https://www.enaminestore.com/catalog/EN300-165246
Availability details for
Quantity
  • Availability
  • Add to Cart