ENA423205169 Enamine

2-(ethylamino)-2-phenylethan-1-ol

  • MDL number MFCD09035099
  • Cas Number : 24417-07-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-165596

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-165596
Availability details for
Quantity
  • Availability
  • Add to Cart