ENA423205170 Enamine

2-(benzylamino)propan-1-ol

  • MDL number MFCD00021997

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-165597

Safety Information

msds https://www.enaminestore.com/catalog/EN300-165597
Availability details for
Quantity
  • Availability
  • Add to Cart