ENA423205401 Enamine

4-bromo-2-chloro-N-(propan-2-yl)aniline

  • MDL number MFCD11643620
  • Cas Number : 1098353-63-9
  • Molecular Weight : 248.55

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2-chloro-N-isopropylaniline
Inchi Code 1S/C9H11BrClN/c1-6(2)12-9-4-3-7(10)5-8(9)11/h3-6,12H,1-2H3
Inchi Key MMEBZTPZVFYIEJ-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-165830

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-165830
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart