ENA423205416 Enamine

N-ethyl-4-methyl-2-nitroaniline

  • MDL number MFCD07437779
  • Cas Number : 37637-58-4
  • Molecular Weight : 180.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-165845

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-165845
Availability details for
Quantity
  • Availability
  • Add to Cart