ENA423206059 Enamine

7-fluoro-4-(trifluoromethyl)-1H-indole

  • MDL number MFCD12405152
  • Cas Number : 1167055-51-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-166489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-166489
Availability details for
Quantity
  • Availability
  • Add to Cart