ENA423206201 Enamine

N-[(1-methyl-1H-pyrazol-5-yl)methyl]aniline

  • MDL number MFCD04968521
  • Cas Number : 1006352-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-166633

Safety Information

msds https://www.enaminestore.com/catalog/EN300-166633
Availability details for
Quantity
  • Availability
  • Add to Cart