ENA423206210 Enamine

4-fluoro-N-[(1-methyl-1H-pyrazol-5-yl)methyl]aniline

  • MDL number MFCD04968536
  • Cas Number : 1006493-59-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-166642

Safety Information

msds https://www.enaminestore.com/catalog/EN300-166642
Availability details for
Quantity
  • Availability
  • Add to Cart