ENA423206240 Enamine

3-fluoro-N-[(3-methyl-1H-pyrazol-4-yl)methyl]aniline

  • MDL number MFCD12112741
  • Cas Number : 1157108-45-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-166672

Safety Information

msds https://www.enaminestore.com/catalog/EN300-166672
Availability details for
Quantity
  • Availability
  • Add to Cart