ENA423206247 Enamine

2-fluoro-N-[(3-methyl-1H-pyrazol-4-yl)methyl]aniline

  • MDL number MFCD12112751
  • Cas Number : 1156888-70-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-166679

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-166679
Availability details for
Quantity
  • Availability
  • Add to Cart