ENA423206765 Enamine

1-methyl-N-(propan-2-yl)-1H-pyrazol-3-amine

  • MDL number MFCD12156902
  • Cas Number : 1178319-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-167198

Safety Information

msds https://www.enaminestore.com/catalog/EN300-167198
Availability details for
Quantity
  • Availability
  • Add to Cart