ENA423206784 Enamine

tert-butyl[(4-methylphenyl)methyl]amine

  • MDL number MFCD01534063
  • Cas Number : 55980-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-167217

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-167217
pictograms GHS05
Availability details for
Quantity
  • Availability
  • Add to Cart