ENA423206852 Enamine

(2-methylpropyl)[(5-methylthiophen-2-yl)methyl]amine

  • MDL number MFCD04488628
  • Cas Number : 774547-45-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-167285

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-167285
Availability details for
Quantity
  • Availability
  • Add to Cart