ENA423206859 Enamine

tert-butyl[(4-methoxyphenyl)methyl]amine

  • MDL number MFCD01427015
  • Cas Number : 22675-83-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-167292

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-167292
Availability details for
Quantity
  • Availability
  • Add to Cart