ENA423206891 Enamine

N-[(4-methylphenyl)methyl]aniline

  • MDL number MFCD00026015
  • Cas Number : 15818-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-167324

Safety Information

msds https://www.enaminestore.com/catalog/EN300-167324
Availability details for
Quantity
  • Availability
  • Add to Cart