ENA423206893 Enamine

N-benzyl-4-fluoroaniline

  • MDL number MFCD00459115
  • Cas Number : 370-77-4
  • Molecular Weight : 201.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-167326

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-167326
Availability details for
Quantity
  • Availability
  • Add to Cart