ENA423207551 Enamine

3-chloro-N-(1H-imidazol-2-ylmethyl)-2-methylaniline

  • MDL number MFCD14693390
  • Cas Number : 913713-83-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168094

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168094
Availability details for
Quantity
  • Availability
  • Add to Cart