ENA423207596 Enamine

N-(1H-imidazol-2-ylmethyl)-4-methylaniline

  • MDL number MFCD08668358
  • Cas Number : 660405-11-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168139

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168139
Availability details for
Quantity
  • Availability
  • Add to Cart