ENA423207611 Enamine

2-fluoro-N-(1H-imidazol-2-ylmethyl)-4-methylaniline

  • MDL number MFCD14693507
  • Cas Number : 1250211-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168154

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168154
Availability details for
Quantity
  • Availability
  • Add to Cart