ENA423207613 Enamine

2-fluoro-N-(1H-imidazol-2-ylmethyl)-5-methylaniline

  • MDL number MFCD14693511
  • Cas Number : 1251329-00-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168156

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168156
Availability details for
Quantity
  • Availability
  • Add to Cart