ENA423207654 Enamine

3-fluoro-N-(1H-imidazol-2-ylmethyl)-2-methylaniline

  • MDL number MFCD16066080
  • Cas Number : 1248782-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168197

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168197
Availability details for
Quantity
  • Availability
  • Add to Cart