ENA423207908 Enamine

3-{[(4-methoxyphenyl)methyl]amino}propan-1-ol

  • MDL number MFCD05863661
  • Cas Number : 91340-37-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-168473

Safety Information

msds https://www.enaminestore.com/catalog/EN300-168473
Availability details for
Quantity
  • Availability
  • Add to Cart